(2,6-Dibromo-4-fluorophenyl)methanol ≥97% research grade
Supplier: AOBChem USA
|
Warning
|
32544-1G
32544-25G
32544-500MG
32544-5G
76887-456EA
379.81
USD
76887-456
76887-458
76887-460
76887-462
(2,6-Dibromo-4-fluorophenyl)methanol ≥97% research grade
(2,6-Dibromo-4-fluorophenyl)methanol
Formula:
C₇H₅Br₂FO Storage Temperature: Refrigerator |
MDL Number:
MFCD28717473 CAS Number: 1346674-69-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...