1-Bromo-5-chloro-2,4-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

32333-25G 32333-5G
76887-434EA 240.83 USD
76887-434 76887-436
1-Bromo-5-chloro-2,4-dimethoxybenzene ≥97%
1-Bromo-5-chloro-2,4-dimethoxybenzene
Formula: C₈H₈BrClO₂
MW: 251.505 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30529807
CAS Number: 864745-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR