(6-Chloro-2-methylpyridin-3-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

32299-1G 32299-25G 32299-500MG 32299-5G
76887-422EA 231.28 USD
76887-422 76887-424 76887-426 76887-428
(6-Chloro-2-methylpyridin-3-yl)methanol ≥97%
(6-Chloro-2-methylpyridin-3-yl)methanol
Formula: C₇H₈ClNO
MW: 157.598 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13689158
CAS Number: 1093879-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR