4,4,5,5-Tetramethyl-2-(2,4,6-trimethylphenyl)-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

32182-25G 32182-5G
76887-416EA 403.15 USD
76887-416 76887-418
4,4,5,5-Tetramethyl-2-(2,4,6-trimethylphenyl)-1,3,2-dioxaborolane ≥97%
4,4,5,5-Tetramethyl-2-(2,4,6-trimethylphenyl)-1,3,2-dioxaborolane
Formula: C₁₅H₂₃BO₂
Storage Temperature: Ambient
MDL Number: MFCD22414456
CAS Number: 171364-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR