5-Bromo-2-fluoro-4-iodotoluene ≥97%

Supplier: AOBChem USA

Warning

32138-25G 32138-5G
76887-408EA 488.02 USD
76887-408 76887-410
5-Bromo-2-fluoro-4-iodotoluene ≥97%
5-Bromo-2-fluoro-4-iodotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrFI
MW: 314.92 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00143260
CAS Number: 202865-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR