1-(2-Chloro-3-fluoropyridin-4-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

32091-1G 32091-25G 32091-500MG 32091-5G
76887-394EA 290.69 USD
76887-394 76887-396 76887-398 76887-400
1-(2-Chloro-3-fluoropyridin-4-yl)ethanone ≥97%
1-(2-Chloro-3-fluoropyridin-4-yl)ethanone
Formula: C₇H₅ClFNO
Storage Temperature: Refrigerator
MDL Number: MFCD18257921
CAS Number: 1236770-00-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR