(2-(Benzyloxy)pyridin-4-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

32014-1G 32014-500MG 32014-5G
76887-374EA 832.81 USD
76887-374 76887-376 76887-378
(2-(Benzyloxy)pyridin-4-yl)methanol ≥97%
(2-(Benzyloxy)pyridin-4-yl)methanol
Formula: C₁₃H₁₃NO₂
MW: 215.248 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14606380
CAS Number: 156367-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR