(2-Chloro-6-methoxypyridin-3-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

31885-1G 31885-25G 31885-500MG 31885-5G
76887-338EA 138.98 USD
76887-338 76887-340 76887-342 76887-344
(2-Chloro-6-methoxypyridin-3-yl)methanol ≥97%
(2-Chloro-6-methoxypyridin-3-yl)methanol
Formula: C₇H₈ClNO₂
MW: 173.597 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16606953
CAS Number: 1228898-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR