1,3-Difluoro-5-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

31746-1G 31746-25G 31746-5G 31746-500MG
76887-324EA 108.22 USD
76887-324 76887-326 76887-328 77650-016
1,3-Difluoro-5-isopropoxybenzene ≥97%
1,3-Difluoro-5-isopropoxybenzene
Formula: C₉H₁₀F₂O
MW: 172.172 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28363314
CAS Number: 1369849-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR