1-(3,5-Dichloropyridin-4-yl)ethanol ≥97%

Supplier: AOBChem USA

Warning

31602-1G 31602-25G 31602-5G
76887-296EA 99.73 USD
76887-296 76887-298 76887-300
1-(3,5-Dichloropyridin-4-yl)ethanol ≥97%
1-(3,5-Dichloropyridin-4-yl)ethanol

Precaución: for Research Use Only

Formula: C₇H₇Cl₂NO
MW: 192.043 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23163275
CAS Number: 1254473-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR