2,3-Difluoro-4-methylbenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

31497-25G 31497-5G
76887-286EA 307.67 USD
76887-286 76887-288
2,3-Difluoro-4-methylbenzyl alcohol ≥97%
2,3-Difluoro-4-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₈F₂O
Storage Temperature: Ambient
MDL Number: MFCD01631646
CAS Number: 252004-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR