2,3-Difluoro-4-(phenylmethoxy)benzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

31272-1G 31272-25G 31272-500MG 31272-5G
76887-212EA 213.25 USD
76887-212 76887-214 76887-216 76887-218
2,3-Difluoro-4-(phenylmethoxy)benzyl alcohol ≥97%
2,3-Difluoro-4-(phenylmethoxy)benzyl alcohol
Formula: C₁₄H₁₂F₂O₂
MW: 250.241 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29088284
CAS Number: 1637386-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR