1-Bromo-4-Chloro-3-Fluoro-2-(Methylsulfanyl)Benzene ≥97%

Supplier: AOBChem USA

Warning

31258-1G 31258-25G 31258-5G
76887-200EA 62.6 USD
76887-200 76887-202 76887-204
1-Bromo-4-Chloro-3-Fluoro-2-(Methylsulfanyl)Benzene ≥97%
1-Bromo-4-Chloro-3-Fluoro-2-(Methylsulfanyl)Benzene
Formula: C₇H₅BrClFS
MW: 255.535 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28141969
CAS Number: 1628524-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR