2,4-Dichloro-3-fluorobenzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

31248-1G 31248-25G 31248-5G
76887-170EA 62.6 USD
76887-170 76887-172 76887-174
2,4-Dichloro-3-fluorobenzaldehyde ≥97% research grade
2,4-Dichloro-3-fluorobenzaldehyde
Formula: C₇H₃Cl₂FO
Storage Temperature: Ambient
MDL Number: MFCD26407306
CAS Number: 1785621-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR