2,3-Dichloro-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

31239-1G 31239-25G 31239-5G
76887-158EA 81.69 USD
76887-158 76887-160 76887-162
2,3-Dichloro-4-methylbenzaldehyde ≥97%
2,3-Dichloro-4-methylbenzaldehyde
Formula: C₈H₆Cl₂O
Storage Temperature: Refrigerator
MDL Number: MFCD18917081
CAS Number: 170879-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR