2-(2,3-Dichloro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

31236-1G 31236-25G 31236-500MG 31236-5G
76887-142EA 409.51 USD
76887-142 76887-144 76887-146 76887-148
2-(2,3-Dichloro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(2,3-Dichloro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₃H₁₇BCl₂O₂
MW: 286.99 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29088283
CAS Number: 2121512-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR