1-Bromo-5-chloro-2-fluoro-4-ethoxybenzene ≥95%

Supplier: AOBChem USA

Warning

31199-1G 31199-25G 31199-500MG 31199-5G
76887-126EA 266.29 USD
76887-126 76887-128 76887-130 76887-132
1-Bromo-5-chloro-2-fluoro-4-ethoxybenzene ≥95%
1-Bromo-5-chloro-2-fluoro-4-ethoxybenzene
Formula: C₈H₇BrClFO
MW: 253.496 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29088273
CAS Number: 1865026-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR