(3-Chloro-6-fluoro-2-methoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

31176-1G 31176-25G 31176-500MG 31176-5G
76887-110EA 171.87 USD
76887-110 76887-112 76887-114 76887-116
(3-Chloro-6-fluoro-2-methoxyphenyl)methanol ≥97%
(3-Chloro-6-fluoro-2-methoxyphenyl)methanol
Formula: C₈H₈ClFO₂
MW: 190.599 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28523085
CAS Number: 1780183-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR