1,2-Dimethoxy-3-(methoxymethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

31123-25G 31123-5G 31123-1G
76887-100EA 616.39 USD
76887-100 76887-102 77637-968
1,2-Dimethoxy-3-(methoxymethoxy)benzene ≥95%
1,2-Dimethoxy-3-(methoxymethoxy)benzene
Formula: C₁₀H₁₄O₄
MW: 198.216 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22123710
CAS Number: 154045-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR