[2-Fluoro-5-(Methylsulfanyl)Phenyl]Methanol ≥97%

Supplier: AOBChem USA

Warning

30981-1G 30981-25G 30981-500MG 30981-5G
76887-034EA 286.45 USD
76887-034 76887-036 76887-038 76887-040
[2-Fluoro-5-(Methylsulfanyl)Phenyl]Methanol ≥97%
[2-Fluoro-5-(Methylsulfanyl)Phenyl]Methanol
Formula: C₈H₉FOS
Storage Temperature: Refrigerator
MDL Number: MFCD34564019
CAS Number: 1510440-59-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR