2,4-Dichloro-5-iodobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

30877-1G 30877-25G 30877-500MG 30877-5G
76887-018EA 224.92 USD
76887-018 76887-020 76887-022 76887-024
2,4-Dichloro-5-iodobenzoic acid ≥97%
2,4-Dichloro-5-iodobenzoic acid
Formula: C₇H₃Cl₂IO₂
Storage Temperature: Ambient
MDL Number: MFCD24107210
CAS Number: 959762-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR