2-(Ethoxycarbonyl)thiophen-3-ylboronic acid ≥97%

Supplier: AOBChem USA

Warning

30856-1G 30856-25G 30856-500MG 30856-5G
76887-002EA 231.28 USD
76887-002 76887-004 76887-006 76887-008
2-(Ethoxycarbonyl)thiophen-3-ylboronic acid ≥97%
2-(Ethoxycarbonyl)thiophen-3-ylboronic acid
Formula: C₇H₉BO₄S
MW: 200.02 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18383157
CAS Number: 632325-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR