1-(5-Hydroxy-2-methylphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

30815-1G 30815-25G 30815-500MG 30815-5G
76886-986EA 439.22 USD
76886-986 76886-988 76886-990 76886-992
1-(5-Hydroxy-2-methylphenyl)ethanone ≥97%
1-(5-Hydroxy-2-methylphenyl)ethanone
Formula: C₉H₁₀O₂
MW: 150.174 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18397235
CAS Number: 40180-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR