2,3,4-Trichloropyridine ≥97%

Supplier: AOBChem USA

Warning

30801-25G 30801-5G
76886-978EA 630.18 USD
76886-978 76886-980
2,3,4-Trichloropyridine ≥97%
2,3,4-Trichloropyridine

Precaución: for Research Use Only

Formula: C₅H₂Cl₃N
Storage Temperature: Refrigerator
MDL Number: MFCD22576127
CAS Number: 55934-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR