2-(Benzyloxy)nicotinic acid ≥97%

Supplier: AOBChem USA

Warning

30536-1G 30536-25G 30536-500MG 30536-5G
76886-942EA 201.58 USD
76886-942 76886-944 76886-946 76886-948
2-(Benzyloxy)nicotinic acid ≥97%
2-(Benzyloxy)nicotinic acid
Formula: C₁₃H₁₁NO₃
MW: 229.231 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00442989
CAS Number: 14178-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR