1-(6-Bromopyridin-3-yl)ethan-1-ol ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-(6-Bromopyridin-3-yl)ethanol

30522-25G 30522-5G
76886-930EA 595.17 USD
76886-930 76886-932
1-(6-Bromopyridin-3-yl)ethan-1-ol ≥95%
1-(6-Bromopyridin-3-yl)ethan-1-ol
Formula: C₇H₈BrNO
MW: 202.049 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20278034
CAS Number: 139042-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR