(2-Bromo-6-fluoro-3-methylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

30418-1G 30418-25G 30418-5G
76886-904EA 64.72 USD
76886-904 76886-906 76886-908
(2-Bromo-6-fluoro-3-methylphenyl)methanol ≥97%
(2-Bromo-6-fluoro-3-methylphenyl)methanol
Formula: C₈H₈BrFO
MW: 219.051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22205827
CAS Number: 1375069-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR