2,3-Difluoro-4-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

30302-25G 30302-5G
76886-888EA 219.61 USD
76886-888 76886-890
2,3-Difluoro-4-methylbenzamide ≥97%
2,3-Difluoro-4-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₇F₂NO
MW: 171.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631633
CAS Number: 261763-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR