(5-Bromo-2-methoxypyridin-3-yl)methanol ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-2-methoxy-3-pyridinemethanol

30226-1G 30226-25G 30226-5G
76886-876EA 96.55 USD
76886-876 76886-878 76886-880
(5-Bromo-2-methoxypyridin-3-yl)methanol ≥97% research grade
(5-Bromo-2-methoxypyridin-3-yl)methanol
Formula: C₇H₈BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD11840327
CAS Number: 351410-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR