(3-Chloropyridin-2-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

30084-1G 30084-25G 30084-5G
76886-868EA 109.28 USD
76886-868 76886-870 76886-872
(3-Chloropyridin-2-yl)methanol ≥97%
(3-Chloropyridin-2-yl)methanol
Formula: C₆H₆ClNO
Storage Temperature: Refrigerator
MDL Number: MFCD11110226
CAS Number: 60588-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR