2-(6-Fluoropyridin-2-yl)acetic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
30015-1G
30015-25G
30015-500MG
30015-5G
76886-852EA
439.22
USD
76886-852
76886-854
76886-856
76886-858
2-(6-Fluoropyridin-2-yl)acetic acid ≥97%
2-(6-Fluoropyridin-2-yl)acetic acid
Formula:
C₇H₆FNO₂ MW: 155.126 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09925118 CAS Number: 1000517-25-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...