(5-Bromo-2-chloropyridin-4-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-2-chloro-4-(hydroxymethyl)pyridine

29972-1G 29972-25G 29972-500MG 29972-5G
76886-842EA 153.84 USD
76886-842 76886-844 76886-846 76886-848
(5-Bromo-2-chloropyridin-4-yl)methanol ≥97%
(5-Bromo-2-chloropyridin-4-yl)methanol
Formula: C₆H₅BrClNO
MW: 222.47 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11847347
CAS Number: 1211531-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR