5-Bromo-3-chloro-2-fluorobenzyl alcohol ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (5-Bromo-3-chloro-2-fluorophenyl)methanol

29843-1G 29843-25G 29843-500MG 29843-5G
76886-826EA 201.58 USD
76886-826 76886-828 76886-830 76886-832
5-Bromo-3-chloro-2-fluorobenzyl alcohol ≥95%
5-Bromo-3-chloro-2-fluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅BrClFO
MW: 239.469 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29036608
CAS Number: 1514304-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR