2,3-Dichloro-5-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,3-Dichloropyridine-5-boronic acid

29784-1G 29784-25G 29784-5G
76886-814EA 84.88 USD
76886-814 76886-816 76886-818
2,3-Dichloro-5-pyridineboronic acid ≥97%
2,3-Dichloro-5-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₅H₄BCl₂NO₂
MW: 191.81 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08277308
CAS Number: 1072944-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR