1-Bromo-2-methoxy-4,5-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

29677-1G 29677-25G 29677-500MG 29677-5G
76886-802EA 165.51 USD
76886-802 76886-804 76886-806 76886-808
1-Bromo-2-methoxy-4,5-dimethylbenzene ≥97%
1-Bromo-2-methoxy-4,5-dimethylbenzene
Formula: C₉H₁₁BrO
MW: 215.087 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16309015
CAS Number: 33500-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR