1-(2,2-Diethoxyethoxy)-4-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

29612-1G 29612-25G 29612-500MG 29612-5G
76886-772EA 345.86 USD
76886-772 76886-774 76886-776 76886-778
1-(2,2-Diethoxyethoxy)-4-fluorobenzene ≥97%
1-(2,2-Diethoxyethoxy)-4-fluorobenzene
Formula: C₁₂H₁₇FO₃
Storage Temperature: Refrigerator
MDL Number: MFCD18848554
CAS Number: 59769-37-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR