1-(4-Bromobenzoyl)-4-methylpiperazine ≥95%

Supplier: AOBChem USA

Warning

29354-25G 29354-5G
76886-706EA 453.01 USD
76886-706 76886-708
1-(4-Bromobenzoyl)-4-methylpiperazine ≥95%
1-(4-Bromobenzoyl)-4-methylpiperazine
Formula: C₁₂H₁₅BrN₂O
MW: 283.164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00448836
CAS Number: 349395-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR