2-(Benzyloxy)-4-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

29186-1G 29186-25G 29186-500MG 29186-5G
76886-668EA 188.85 USD
76886-668 76886-670 76886-672 76886-674
2-(Benzyloxy)-4-methoxybenzoic acid ≥97%
2-(Benzyloxy)-4-methoxybenzoic acid
Formula: C₁₅H₁₄O₄
MW: 258.269 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09719635
CAS Number: 13618-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR