2-(Benzyloxy)-4-methoxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

29181-1G 29181-500MG 29181-5G
76886-656EA 528.33 USD
76886-656 76886-658 76886-660
2-(Benzyloxy)-4-methoxyphenylboronic acid pinacol ester ≥97%
Formula: C₂₀H₂₅BO₄
Storage Temperature: Refrigerator
MDL Number: MFCD28806020
CAS Number: 2121511-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR