1-Bromo-4-difluoromethoxy-2-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

29170-1G 29170-25G 29170-500MG 29170-5G
76886-642EA 122.01 USD
76886-642 76886-644 76886-646 76886-648
1-Bromo-4-difluoromethoxy-2-fluorobenzene ≥95%
1-Bromo-4-difluoromethoxy-2-fluorobenzene

Precaución: for Research Use Only

Formula: C₇H₄BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD18390897
CAS Number: 1242258-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR