4-(Difluoromethoxy)benzeneboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (4-(Difluoromethoxy)phenyl)boronic acid

29157-1G 29157-25G 29157-500MG 29157-5G
76886-626EA 138.98 USD
76886-626 76886-628 76886-630 76886-632
4-(Difluoromethoxy)benzeneboronic acid ≥95%
4-(Difluoromethoxy)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BF₂O₃
MW: 187.936 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09864667
CAS Number: 688810-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR