1-(3-Bromophenyl)cyclopentan-1-ol ≥95%

Supplier: AOBChem USA

Warning

29000-1G 29000-25G 29000-5G
76886-600EA 63.66 USD
76886-600 76886-602 76886-604
1-(3-Bromophenyl)cyclopentan-1-ol ≥95%
1-(3-Bromophenyl)cyclopentan-1-ol
Formula: C₁₁H₁₃BrO
MW: 241.124 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23708970
CAS Number: 210826-69-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR