1-Methoxy-4-(methoxymethoxy)benzene 95%

Supplier: AOBChem USA

Warning

28882-25G 28882-5G 28882-1G
76886-588EA 616.39 USD
76886-588 76886-590 77637-816
1-Methoxy-4-(methoxymethoxy)benzene 95%
1-Methoxy-4-(methoxymethoxy)benzene
Formula: C₉H₁₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD27948192
CAS Number: 25458-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR