2-(Methoxymethoxy)-3-methylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

28721-1G 28721-25G 28721-500MG 28721-5G
76886-544EA 358.59 USD
76886-544 76886-546 76886-548 76886-550
2-(Methoxymethoxy)-3-methylbenzaldehyde ≥95%
2-(Methoxymethoxy)-3-methylbenzaldehyde
Formula: C₁₀H₁₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD24674140
CAS Number: 55359-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR