1-Bromo-3-iodo-5-isopropylbenzene ≥97%

Supplier: AOBChem USA

Warning

28691-1G 28691-25G 28691-5G 28691-500MG
76886-530EA 79.57 USD
76886-530 76886-532 76886-534 77645-728
1-Bromo-3-iodo-5-isopropylbenzene ≥97%
1-Bromo-3-iodo-5-isopropylbenzene
Formula: C₉H₁₀BrI
MW: 324.984 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28360703
CAS Number: 1369835-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR