1-(4-Bromophenyl)-2,5-dimethylpyrrole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-(4-Bromophenyl)-2,5-dimethyl-1H-pyrrole

28677-25G 28677-5G
76886-518EA 268.41 USD
76886-518 76886-520
1-(4-Bromophenyl)-2,5-dimethylpyrrole ≥97%
1-(4-Bromophenyl)-2,5-dimethylpyrrole

Precaución: for Research Use Only

Formula: C₁₂H₁₂BrN
Storage Temperature: Ambient
MDL Number: MFCD00277042
CAS Number: 5044-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR