1-(4-Bromophenyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

28613-1G 28613-25G 28613-500MG 28613-5G
76886-502EA 147.47 USD
76886-502 76886-504 76886-506 76886-508
1-(4-Bromophenyl)cyclohexanol ≥95%
1-(4-Bromophenyl)cyclohexanol
Formula: C₁₂H₁₅BrO
Storage Temperature: Refrigerator
MDL Number: MFCD18205680
CAS Number: 19920-80-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR