(4-Cyclobutylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

28580-1G 28580-25G 28580-500MG 28580-5G
76886-486EA 257.8 USD
76886-486 76886-488 76886-490 76886-492
(4-Cyclobutylphenyl)methanol ≥97%
(4-Cyclobutylphenyl)methanol
Formula: C₁₁H₁₄O
Storage Temperature: Refrigerator
MDL Number: MFCD19440004
CAS Number: 1379071-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR