1-Bromo-4-cyclobutylbenzene ≥97%

Supplier: AOBChem USA

Warning

28512-1G 28512-25G 28512-500MG 28512-5G
76886-454EA 155.96 USD
76886-454 76886-456 76886-458 76886-460
1-Bromo-4-cyclobutylbenzene ≥97%
1-Bromo-4-cyclobutylbenzene
Formula: C₁₀H₁₁Br
Storage Temperature: Refrigerator
MDL Number: MFCD19439999
CAS Number: 39868-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR