5,6-Difluorosalicylic acid ≥95%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2,3-Difluoro-6-hydroxybenzoic acid
28510-1G
28510-25G
28510-500MG
28510-5G
76886-438EA
162.32
USD
76886-438
76886-440
76886-442
76886-444
5,6-Difluorosalicylic acid ≥95%
5,6-Difluorosalicylic acid
Formula:
C₇H₄F₂O₃ MW: 174.1017 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD07368161 CAS Number: 749230-47-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...